Learn More
Axygen™ Maxymum Recovery™ 300 μL Universal Pipetter Tips
Shop All Axygen Scientific ProductsDescription
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors, compatible with most popular brands of pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
- These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
- Nonpyrogenic
- Fine-Point
- Maxymum Recovery technology for ultra-low retention and high yield
Order Info
- Bulk: 1000 tips/pack, 240 packs/case
- Racked: 96 tips/rack, 10 racks/pack, 5 packs/case
Specifications
Specifications
| Volume (Metric) | 1 to 300 μL |
| Sterility | Non-sterile |
| Color | Clear |
| Clean Claims | DNase, RNase, ATP, and Endotoxin Free |
| Format | Rack |
| Tip Style | Low Retention, Universal |
| Packaging | Hinged rack |
| For Use With (Equipment) | Universal Fit |
| Product Line | Maxymum Recovery |
| Material | Polypropylene |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.