Learn More
Axygen™ 200 μL Universal Pipetter Tips: 200 μL, Beveled, Reference
Shop All Axygen Scientific ProductsDescription
Axygen™ Universal Tips are designed to fit a wide variety of single and multi-channel pipettors. These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free. They are available in Maxymum Recovery, beveled, nonsterile, sterile, non-filtered, and other features for specialized applications. Flexible walls and a series of internal sealing rings ensure a secure fit with less force required to load and eject.
(This is the suggested replacement for T-205-WB-C-L)
- These polypropylene tips are nonpyrogenic and certified to be RNase-/DNase-free.
- Nonpyrogenic
- Wide-Bore
Order Info
Available in bulk, racked or stack rack formats.
Specifications
Specifications
| Description | Graduated pipet tip |
| Volume (Metric) | 1 to 200 μL |
| Filtered | Non-filtered |
| Sterility | Non-sterile |
| Color | Clear |
| Clean Claims | DNase, RNase Free |
| Format | Refill |
| Tip Style | Beveled, Low Retention |
| Packaging | 96 Tips/Rack, 5 Racks/Stack, 10 Stacks/Cs |
| Disposable | Single-use |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.