Learn More
Axitinib, TRC
Specifications
Specifications
| Chemical Name or Material | Axitinib |
| CAS | 319460-85-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C22 H18 N4 O S |
| Quantity | 100 mg |
| Synonym | Benzamide, N-methyl-2-[[3-[(1E)-2-(2-pyridinyl)ethenyl]-1H-indazol-6-yl]thio]-, N-Methyl-2-[[3-[(1E)-2-(2-pyridinyl)ethenyl]-1H-indazol-6-yl]thio]benzamide, AG 013736, Axitinib, Inlyta, Benzamide, N-methyl-2-[[3-[(1E)-2-(2-pyridinyl)ethenyl]-1H-indazol-6-yl]thio]-, N-Methyl-2-[[3-[(1E)-2-(2-pyridinyl)ethenyl]-1H-indazol-6-yl]thio]benzamide, AG 013736, Axitinib, Inlyta |
| InChI Formula | InChI=1S/C22H18N4OS/c1-23-22(27)18-7-2-3-8-21(18)28-16-10-11-17-19(25-26-20(17)14-16)12-9-15-6-4-5-13-24-15/h2-14H,1H3,(H,23,27)(H,25,26)/b12-9+ |
| SMILES | CNC(=O)c1ccccc1Sc2ccc3c(\C=C\c4ccccn4)n[nH]c3c2 |
| IUPAC Name | N-methyl-2-[[3-[(E)-2-pyridin-2-ylethenyl]-1H-indazol-6-yl]sulfanyl]benzamide |
| Molecular Weight (g/mol) | 386.47 |
| Show More |