missing translation for 'onlineSavingsMsg'
Learn More

AX-15836, MedChemExpress

Product Code. 16446337
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16446337

Brand: Medchem Express HY101846

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AX-15836 is a potent and selective ERK5 inhibitor with an IC50 of 8 nM.

AX-15836 is a potent and selective ERK5 inhibitor with an IC50 of 8 nM.

Specifications

Chemical Name or Material AX-15836
CAS 2035509-96-5
Color Off-White
Purity Grade Notes Research
Molecular Formula C32H40N8O5S
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (154.14 mM; Need ultrasonic)
SMILES O=C1N(C)C2=CN=C(NC3=CC=C(C(N4CCC(N5CCN(C)CC5)CC4)=O)C=C3OCC)N=C2N(S(=O)(C)=O)C6=CC=CC=C16
Molecular Weight (g/mol) 648.78
Formula Weight 648.78
Percent Purity 99.68%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.