missing translation for 'onlineSavingsMsg'
Learn More

Avitinib maleate, MedChemExpress

Product Code. 16437928
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16437928

Brand: Medchem Express HY19816A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Avitinib (Abivertinib) maleate is a pyrrolopyrimidine-based irreversible epidermal growth factor receptor (EGFR) inhibitor with an IC50 of 7.68 nM.

Avitinib (Abivertinib) maleate is a pyrrolopyrimidine-based irreversible epidermal growth factor receptor (EGFR) inhibitor with an IC50 of 7.68 nM.

Specifications

Chemical Name or Material Avitinib maleate
CAS 1557268-88-8
Color Yellow
Purity Grade Notes Research
Molecular Formula C30H30FN7O6
Quantity 10 mg
Synonym Abivertinib maleateAC0010 maleate
Solubility Information DMSO : ≥ 100 mg/mL (165.67 mM) ∣H2O : < 0.1 mg/mL (ultrasonic;warming;heat to 60°C) (insoluble)
SMILES O=C(O)/C=C\C(O)=O.O=C(C=C)NC1=CC=CC(OC2=NC(NC3=CC(F)=C(N4CCN(C)CC4)C=C3)=NC5=C2C=CN5)=C1
Molecular Weight (g/mol) 603.6
Formula Weight 603.6
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.