missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Avian Influenza NS2 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0396
Additional Details : Weight : 0.01000kg
Description
Avian Influenza Nonstructural Protein 2 Synthetic Peptide, PEP-0396, from Invitrogen.
This peptide corresponds to 14 amino acids near the center of avian influenza NS2.Specifications
Virus | |
Avian Influenza NS2 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
0.02% sodium azide | |
50 μg | |
PBS |
purified | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
No | |
Peptide | |
RUO - research use only |