missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AVE3085, MedChemExpress
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mg
5 mg
10 mg
20 mg
missing translation for 'unitSize'
10mg
1mg
20mg
5mg
Description
AVE3085 is a potent endothelial nitric oxide synthase enhancer, used for cardiovascular disease treatment.
Specifications
Specifications
| Chemical Name or Material | AVE3085 |
| CAS | 450348-85-3 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C17H13F2NO3 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 250 mg/mL (787.92 mM; Need ultrasonic) |
| SMILES | O=C(C1=CC=C(OC(F)(F)O2)C2=C1)NC3CC4=C(C=CC=C4)C3 |
| Molecular Weight (g/mol) | 317.29 |
| Formula Weight | 317.29 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction