Learn More
ABEBA™ Autoclave Clog OB
Product Code. 16348231
Change view
Click to view available options
Color:
Blue
Green
White
Unit Size:
1 pair
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
16348231 White 1 pair
16358231 Blue 1 pair
16368231 Green 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16348231 Supplier ABEBA™ Supplier No. 3960035
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
ABEBA™ Autoclave Clog OB is designed for sterile environments and features a foldable heel strap with a slip resistant outsole.
  • Hygiene, stability, durability
  • Comply to EN ISO 20347 : 2012, OB A E SRC
  • ESD dissipative according to EN 61340 Climate class 1
  • Slip resistant outsole SRC
  • Good cushioning
  • Special sterilizable material (135°C)
  • Foldable heel strap, adjustable
  • Sanitized treatment
Specifications
Buckle Material Autoclavable, sterilizable special material
Certifications/Compliance CE EN ISO 20347:2012 OB A, E, SRC, ESD
Color White
No. of Buckles 1
Size 35
Style Clog
Packaging Quantity 1
Sterility Sterile
Certifications/Compliance (Europe) CE EN ISO 20347:2012, DIN EN 61340
Resistant To Autoclave up to 135°C