Learn More
Advantage-Lab™ Autoclave Basket
Product Code. 15678785
Encompass
Change view
Click to view available options
For Use With (Equipment):
AL02-03-100 Autoclave
AL02-13-100 Autoclave
Dimensions (L x W x H):
400 x 170 x 100 mm
500 x 260 x 200 mm
500 x 260 x 225 mm
Description:
Closed basket
Wire basket
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Dimensions (L x W x H) Description unitSize
15678785 AL02-13-100 Autoclave 500 x 260 x 200 mm Closed basket Each
10263693 AL02-03-100 Autoclave 400 x 170 x 100 mm Wire basket Each
15678845 AL02-13-100 Autoclave 500 x 260 x 225 mm Wire basket -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15678785 Supplier Advantage-Lab™ Supplier No. AL0213151
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Durable container constructed of stainless steel. Advantage-Lab™ Autoclave Basket is a suitable container for a variety of autoclaving needs.
  • Stainless steel construction.
Specifications
Autoclavable Autoclavable
Description Closed basket
For Use With (Equipment) AL02-13-100 Autoclave
Material Stainless Steel
Dimensions (L x W x H) 500 x 260 x 200 mm