missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AURKB (Aurora B) Recombinant Human Protein
The Aurora kinases associate with microtubules during chromosome movement and segregation.
1826.00 € - 7890.00 €
Specifications
Accession Number | NP_004208 |
---|---|
For Use With (Application) | Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology |
Formulation | 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol |
Molecular Weight (g/mol) | 43.4kDa |
Storage Requirements | Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15438723
|
Invitrogen™
PR9210B |
100μg |
1826.00 €
100µg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15458813
|
Invitrogen™
PV6131 |
1mg |
7890.00 €
1mg |
Estimated Shipment: 28-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
AURKB (Aurora B) localizes to microtubules near kinetochores, specifically to the specialized microtubules called K-fibers.
Specifications
NP_004208 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
Aurora Kinase Family | |
Recombinant | |
Baculovirus |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
43.4kDa | |
RUO - research use only | |
AURKB (Aurora B) | |
Human | |
His Tag | |
Frozen |