Learn More
Auristatin F, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Description
Auristatin F is a potent cytotoxin. Auristatin F, a potent microtubule inhibitor and vascular damaging agent (VDA), can be used in antibody-drug conjugates (ADC).
Specifications
Specifications
| Chemical Name or Material | Auristatin F |
| Physical Form | Solid |
| CAS | 163768-50-1 |
| Color | White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C40H67N5O8 |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : 200 mg/mL (268.10 mM; Need ultrasonic) |
| SMILES | CC(C)[C@H](N(C)C)C(N[C@H](C(N([C@@H]([C@@H](C)CC)[C@@H](CC(N1CCC[C@@]1([H])[C@H](OC)[C@@H](C)C(N[C@H](C(O)=O)CC2=CC=CC=C2)=O)=O)OC)C)=O)C(C)C)=O |
| Show More |