missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Auristatin F, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Description
Auristatin F is a potent cytotoxin. Auristatin F, a potent microtubule inhibitor and vascular damaging agent (VDA), can be used in antibody-drug conjugates (ADC).
Specifications
Specifications
| Chemical Name or Material | Auristatin F |
| Physical Form | Solid |
| CAS | 163768-50-1 |
| Color | White |
| Quantity | 1 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C40H67N5O8 |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : 200 mg/mL (268.10 mM; Need ultrasonic) |
| SMILES | CC(C)[C@H](N(C)C)C(N[C@H](C(N([C@@H]([C@@H](C)CC)[C@@H](CC(N1CCC[C@@]1([H])[C@H](OC)[C@@H](C)C(N[C@H](C(O)=O)CC2=CC=CC=C2)=O)=O)OC)C)=O)C(C)C)=O |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction