Learn More
Invitrogen™ Attune™ NxT Focus Bottle
Description
- This is a 1.9-L replacement bottle for the focusing fluid tank on the Attune NxT Acoustic Focusing Cytometer.
- The bottle is designed for easy detachment from the instrument for refill and with more robust fittings to prevent breakage.
- This bottle replaces discontinued Cat. No. 100022155.
Specifications
Specifications
| Content And Storage | 1 Bottle |
| For Use With (Equipment) | Attune NxT |
| Format | Bottle |
| Product Line | FPL |
| Quantity | 1 bottle |
| Shipping Condition | Room temperature |
| Product Type | Flow Cytometer Accessory |