Learn More
Invitrogen™ Attractin Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5100899
Product Code. 16627384
  • 493.00 € / 100µL
Please to purchase this item. Need a web account? Register with us today!
Description
Description
Antibody detects endogenous levels of total ATRN.
Multiple transcript variants encoding different isoforms exist for this gene. One of the isoforms is a membrane-bound protein with sequence similarity to the mouse mahogany protein, a receptor involved in controlling obesity. The other isoform is a secreted protein involved in the initial immune cell clustering during inflammatory responses that may regulate the chemotactic activity of chemokines.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Attractin
Polyclonal
Unconjugated
ATRN
ATRN; atrn {ECO:0000312; Attractin; attractin-2; AW558010; DPPT-L; KIAA0548; mahogany; mahogany homolog; mahogany protein; membrane attractin; mg; MGC126754; Mgca; mKIAA0548; Protein mahogany; protein zitter; RGD:69063}
Rabbit
Affinity chromatography
RUO
11990, 8455
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O75882, Q9WU60
ATRN
A synthesized peptide derived from human ATRN(Accession O75882), corresponding to amino acid residues A83-G133.
100 μL
Primary
Human, Mouse