Learn More
Invitrogen™ Attractin Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5100899
Product Code. 16627384
  • 493.00 € / 100µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Antibody detects endogenous levels of total ATRN.
Multiple transcript variants encoding different isoforms exist for this gene. One of the isoforms is a membrane-bound protein with sequence similarity to the mouse mahogany protein, a receptor involved in controlling obesity. The other isoform is a secreted protein involved in the initial immune cell clustering during inflammatory responses that may regulate the chemotactic activity of chemokines.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Attractin
Polyclonal
Unconjugated
ATRN
ATRN; atrn {ECO:0000312; Attractin; attractin-2; AW558010; DPPT-L; KIAA0548; mahogany; mahogany homolog; mahogany protein; membrane attractin; mg; MGC126754; Mgca; mKIAA0548; Protein mahogany; protein zitter; RGD:69063}
Rabbit
Affinity chromatography
RUO
11990, 8455
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O75882, Q9WU60
ATRN
A synthesized peptide derived from human ATRN(Accession O75882), corresponding to amino acid residues A83-G133.
100 μL
Primary
Human, Mouse