missing translation for 'onlineSavingsMsg'
Learn More

ATP6V1B1 Monoclonal Antibody (OTI3B11), TrueMAB™, OriGene

Product Code. 17527191
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17527191 30 μL 30µL
17507191 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17527191 Supplier Origene Technologies Supplier No. TA502550S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ATP6V1B1 Monoclonal antibody specifically detects ATP6V1B1 in Canine, Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen ATP6V1B1
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3B11
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene POLR2E
Gene Accession No. B0BNE2, P19388, Q80UW8
Gene Alias ATP6B1; ATP6V1B1; ATPase H+ transporting V1 subunit B1; ATPase, H transporting, lysosomal V1 subunit B1; ATPase, H+ transporting, lysosomal (vacuolar proton pump), beta 56/58 kDa, isoform 1; ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B, isoform 1; ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B1; ATPase, H+ transporting, lysosomal 56/58kDa, V1 subunit B1 (Renal tubular acidosis with deafness); ATPase, H+ transporting, lysosomal V1 subunit B1; ATPase, H+ transporting, V1 subunit B, isoform 1; AW208839; D630003L15; D630030L16Rik; D630039P21Rik; endomembrane proton pump 58 kDa subunit; H(+)-transporting two-sector ATPase, 58kD subunit; H+-ATPase beta 1 subunit; lysosomal 56/58kDa; RTA1B; vacuolar H+-ATPase; vacuolar proton pump 3; vacuolar proton pump subunit B 1; vacuolar proton pump, subunit 3; VATB; V-ATPase B1; V-ATPase B1 subunit; V-ATPase subunit B 1; VMA2; VPP3; Vpp-3; V-type proton ATPase subunit B, kidney isoform
Host Species Mouse
Immunogen Full length human recombinant protein of human POLR2E produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5434, 611859, 66420, 690966
Target Species Canine, Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.