missing translation for 'onlineSavingsMsg'
Learn More

ATP6AP1 Antibody [Allophycocyanin], Novus Biologicals Biologicals™

Product Code. 30500729 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30500729 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30500729 Supplier Novus Biologicals Supplier No. NBP335213APC

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATP6AP1 Polyclonal antibody specifically detects ATP6AP1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP6AP1
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate APC
Formulation PBS
Gene Alias 16A, Ac45, ATP6IP1V-ATPase subunit S1, ATP6S1ORF, ATPase, H+ transporting, lysosomal (vacuolar proton pump), subunit 1, ATPase, H+ transporting, lysosomal accessory protein 1, H+ transporting, lysosomal interacting protein 1, H-ATPase subunit, MGC129781, Protein XAP-3, Vacuolar proton pump subunit S1, VATPS1V-ATPase Ac45 subunit, V-type proton ATPase subunit S1, XAP-3, XAP3V-ATPase S1 accessory protein
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 90-390 of human ATP6AP1 (NP_001174.2).,, Sequence:, LLFLQDKLSIEDFTAYGGVFGNKQDSAFSNLENALDLAPSSLVLPAVDWYAVSTLTTYLQEKLGASPLHVDLATLRELKLNASLPALLLIRLPYTASSGLMAPREVLTGNDEVIGQVLSTLKSEDVPYTAALTAVRPSRVARDVAVVAGGLGRQLLQKQPVSPVIHPPVSYNDTAPRILFWAQNFSVAYKDQWEDLTPLTFGVQELNLTGSFWNDSFARLSLTYERLFGTTVTFKFILANRLYPVSARHWFTMERLEVHSNGSVAYFNASQVTGPSIYSFHCEYVSSLSKKGSLLVARTQP
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 537
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.