Learn More
Invitrogen™ ATP5H Monoclonal Antibody (7F9BG1)

Description
For mouse and rat samples, this antibody will only recognize ATP5H in purified mitochondrial samples. Mouse and rat cell lysates and tissue homogenates are not recommended with this antibody. For positive control, use Isolated mitochondria from Human heart, Bovine heart, Rat heart, Mouse heart, and HepG2, Cultured Human embryonic lung-derived fibroblasts (strain MRC5), HeLa cells.
Specifications
Specifications
| Antigen | ATP5H |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 7F9BG1 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | HEPES buffered saline with 0.02% sodium azide |
| Gene | ATP5PD |
| Gene Accession No. | O75947, P13620, P31399, Q9DCX2 |
| Gene Alias | 0610009D10Rik; ATP synthase D chain, mitochondrial; ATP synthase peripheral stalk subunit d; ATP synthase subunit d, mitochondrial; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit d; ATP synthase, H+ transporting, mitochondrial F0 complex, subunit d isoform a; ATP synthase, H+ transporting, mitochondrial F1F0, subunit d; ATP synthase, H+ transporting, mitochondrial Fo complex subunit D; ATP synthase, H+ transporting, mitochondrial Fo complex, subunit d; Atp5h; Atp5jd; Atp5pd; ATPase subunit d; ATPQ; My032; My032 protein; unnamed protein product; zgc:73071 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.