missing translation for 'onlineSavingsMsg'
Learn More

ATP2C1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Product Code. 18498845 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18498845 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18498845 Supplier Novus Biologicals Supplier No. NBP176566AF750

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ATP2C1 Polyclonal antibody specifically detects ATP2C1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATP2C1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias ATPase, Ca++ transporting, type 2C, member 1, BCPM, benign chronic pemphigus (Hailey-Hailey disease), Ca(2+)-sequestering, calcium-transporting ATPase type 2C member 1, EC 3.6.3, HHD, hSPCA1, HUSSY-28, PMR1L, secretory pathway Ca2+/Mn2+ ATPase 1
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human ATP2C1. The immunogen is located within the last 50 amino acids of ATP2C1. Amino Acid Squence: IKKVERSREKIQKHVS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 27032
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.