missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ATP1B1 (Human) Recombinant Protein

Human ATP1B1 full-length ORF ( AAH00006, 1 a.a. - 303 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00000481-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16185024

  • 508.00 € / 25µg
Estimated Shipment: 25-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MARGKAKEEGSWKKFIWNSEKKEFLGRTGGSWFKILLFYVIFYGCLAGIFIGTIQVMLLTISEFKPTYQDRVAPPGLTQIPQIQKTEISFRPNDPKSYEAYVLNIVRFLEKYKDSAQRDDMIFEDCGDVPSEPKERGDFNHERGERKVCRFKLEWLGNCSGLNDETYGYKEGKPCIIIKLNRVLGFKPKPPKNESLETYPVMKYNPNVLPVQCTGKRDEDKDKVGNVEYFGLGNSPGFPLQYYPYYGKLLQPKYLQPLLAVQFTNLTMDTEIRIECKAYGENIGYSEKDRFQGRFDVKIEVKS
Specifications

Specifications

AAH00006
ATPase, Na+/K+ transporting, beta 1 polypeptide
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ATP1B1
GST
481
Wheat germ expression system
25 μg
ATP1B, MGC1798
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions