missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP Synthase beta Polyclonal Antibody

ATP Synthase beta Polyclonal Antibody
Brand: Invitrogen PA596419
This item is not returnable.
View return policy
Description
ATP Synthase beta Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| ATP Synthase beta | |
| Polyclonal | |
| Unconjugated | |
| ATP5F1B | |
| ATP synthase F1 subunit beta; ATP synthase subunit beta, mitochondrial; ATP synthase, H+ transporting mitochondrial F1 complex, alpha subunit; ATP synthase, H+ transporting mitochondrial F1 complex, beta subunit; ATP synthase, H+ transporting, mitochondrial F1 complex, beta polypeptide; Atp5b; ATP5F1B; ATPMB; ATPSB; beta-subunit; epididymis secretory protein Li 271; f1-ATPase beta; F1-ATPase beta-subunit; F-1-ATPase beta-subunit precursor; fj13e04; fj55c09; HEL-S-271; hm:zehn0534; hypothetical protein LOC554135; im:6793121; MGC5231; mitochondrial ATP synthase beta subunit; mitochondrial ATP synthase subunit beta subunit; mitochondrial ATP synthase, H+ transporting F1 complex beta subunit; mitochondrial ATP synthetase, beta subunit; wu:fj13e04; wu:fj38d01; wu:fj55c09; zgc:111961 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 11947, 171374, 506 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 4.54 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| P06576, P10719, P56480 | |
| ATP5F1B | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 230-529 of human ATPB (NP_0016772). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction