Learn More
Atorvastatin-d5 Sodium Salt, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | Atorvastatin-D5 Sodium Salt (N-phenyl-D5) |
| CAS | 2170106-11-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C33 2H5 H29 F N2 O5 . Na |
| Quantity | 1 mg |
| Synonym | (3R,5R)-7-[2-(4-Fluorophenyl)-5-isopropyl-3-phenyl-4-pentadeuteriophenylcarbamoylpyrrol-1-yl]-3,5-dihydroxyheptanoic acid sodium salt, (βR,δR)-2-(p-Fluorophenyl)-β,δ-dihydroxy-5-isopropyl-3-phenyl-4-[(phenyl-d5)carbamoyl]pyrrole-1-heptanoic acid sodium salt, Atorvastatin-d5 sodium salt, Atorvastatin-D5 sodium salt (N-phenyl-D5) |
| InChI Formula | InChI=1S/C33H35FN2O5.Na/c1-21(2)31-30(33(41)35-25-11-7-4-8-12-25)29(22-9-5-3-6-10-22)32(23-13-15-24(34)16-14-23)36(31)18-17-26(37)19-27(38)20-28(39)40;/h3-16,21,26-27,37-38H,17-20H2,1-2H3,(H,35,41)(H,39,40);/q;+1/p-1/t26-,27-;/m1./s1/i4D,7D,8D,11D,12D; |
| SMILES | [Na+].[2H]c1c([2H])c([2H])c(NC(=O)c2c(c3ccccc3)c(c4ccc(F)cc4)n(CC[C@@H](O)C[C@@H](O)CC(=O)[O-])c2C(C)C)c([2H])c1[2H] |
| IUPAC Name | sodium;(3R,5R)-7-[2-(4-fluorophenyl)-4-[(2,3,4,5,6-pentadeuteriophenyl)carbamoyl]-3-phenyl-5-propan-2-ylpyrrol-1-yl]-3,5-dihydroxyheptanoate |
| Molecular Weight (g/mol) | 585.65 |
| Show More |