missing translation for 'onlineSavingsMsg'
Learn More

ATOH7 Polyclonal Antibody

Product Code. 15574702
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15574702 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15574702 Supplier Invitrogen Supplier No. PA541212

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

ATOH7 Polyclonal Antibody

ATOH7 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen ATOH7
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ATOH7
Gene Accession No. Q8N100
Gene Alias ath5; Atoh7; atonal bHLH transcription factor 7; atonal homolog 5; atonal homolog 7; atonal homolog 7 (Drosophila); atonal homolog bHLH transcription factor 7; BHLHA13; Class A basic helix-loop-helix protein 13; hATH5; Helix-loop-helix protein hATH-5; helix-loop-helix protein mATH-5; Helix-loop-helix protein zATH-5; lak; lakritz; Math5; NCRNA; PHPVAR; Protein atonal homolog 5; protein atonal homolog 7; Protein lakritz; RNANC; si:packtrz.1; zATH5
Gene Symbols ATOH7
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human ATOH7 (aa 102-151).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220202
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.