Learn More
ATN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 19243-1-AP-150UL
 View more versions of this product
Product Code. 16828934
  • 410.00 € / 150µL
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
Dentatorubral pallidoluysian atrophy (DRPLA) is a rare neurodegenerative disorder characterized by cerebellar ataxia, myoclonic epilepsy, choreoathetosis, and dementia. The disorder is related to the expansion from 7-35 copies to 49-93 copies of a trinucleotide repeat (CAG/CAA) within this gene. The encoded protein includes a serine repeat and a region of alternating acidic and basic amino acids, as well as the variable glutamine repeat. Alternative splicing results in two transcripts variants that encode the same protein.
Specifications
Specifications
ATN1
Polyclonal
Unconjugated
ATN1
ATN1, atrophin 1, B37, D12S755E, DRPLA, HRS, NOD
Rabbit
Antigen Affinity Chromatography
RUO
13498, 1822
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.23 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O35126, P54259
ATN1
ATN1 Fusion Protein Ag5835
150 μL
Primary
Human, Mouse