missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATG4B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
200.00 € - 351.00 €
Specifications
| Antigen | ATG4B |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18716853
|
Novus Biologicals
NBP2-24709SS |
0.025 mg |
200.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18719133
|
Novus Biologicals
NBP2-24709 |
0.1 mg |
351.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ATG4B Polyclonal specifically detects ATG4B in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ATG4B | |
| Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| APG4 autophagy 4 homolog B, APG4 autophagy 4 homolog B (S. cerevisiae), APG4B, ATG4 autophagy related 4 homolog B (S. cerevisiae), AUTL1MGC1353, AUT-like 1 cysteine endopeptidase, Autophagin-1, Autophagy-related cysteine endopeptidase 1, Autophagy-related protein 4 homolog B, cysteine protease ATG4B, DKFZp586D1822, EC 3.4.22, EC 3.4.22.-, hAPG4B, KIAA0943 | |
| ATG4B | |
| IgG | |
| Protein G purified | |
| The amino acid sequence used as immunogen is 100% homologous in human, mouse, chimpanzee, monkey, rat, dog and orangutan, 94% homologous in opossum and mouse, 88% in zebrafish, pig, and catfish. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Autophagy, Cancer | |
| PBS with 0.05% Sodium Azide | |
| 23192 | |
| A portion of amino acid 350-400 of human APG4B was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title