Learn More
Abnova™ ATF1 Split FISH Probe
Specifications
Specifications
| Content And Storage | Store at 4°C in the dark |
| Format | Tube |
| For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
| Gene | activating transcription factor 1 |
| Gene Alias | EWS-ATF1,FUS/ATF-1,TREB36 |
| Gene Symbol | ATF1 |
| Gene ID (Entrez) | 466 |
| Product Type | DNA/RNA |
| Quantity | 100 μL |
| Form | Liquid |