missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ATAT1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114922
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total C6orf134.
This gene encodes a protein that localizes to clathrin-coated pits, where it acetylates alpha tubulin on lysine 40. This process may be important in microtubule growth, for instance during chemotaxis and the formation of cilium. Alternative splicing results in multiple transcript variants.
Specifications
| ATAT1 | |
| Polyclonal | |
| Unconjugated | |
| ATAT1 | |
| 0610011P08Rik; 2610008K08Rik; 2610110G12Rik; 3110080J08Rik; Acetyltransferase mec-17 homolog; acetyltransferase mec-17 homolog {ECO:0000255; alpha tubulin acetyltransferase 1; alpha-TAT; alpha-TAT {ECO:0000255; alpha-TAT1; alpha-TAT1 {ECO:0000255; alpha-tubulin N-acetyltransferase 1; alpha-tubulin N-acetyltransferase 1 {ECO:0000255; alpha-tubulin N-acetyltransferase; alpha-tubulin N-acetyltransferase 1; Atat1; C23H6orf134; C6orf134; HAMAP-Rule:MF_03130}; hypothetical protein FLJ13158; MEC17; Nbla00487; Novel DUF738 containing protein (2610110G12Rik); RGD1303066; TAT; TAT {ECO:0000255 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 361789, 73242, 79969 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5SQI0, Q6MG11, Q8K341 | |
| ATAT1 | |
| A synthesized peptide derived from human ATAT1(Accession Q5SQI0), corresponding to amino acid residues E229-R279. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction