missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATAD4 / PRR15L Rabbit Polyclonal Antibody, Proteintech
Rabbit Polyclonal Antibody
129.00 € - 349.00 €
Specifications
| Antigen | ATAD4 / PRR15L |
|---|---|
| Concentration | 650 μg/mL |
| Applications | Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30237350
|
Proteintech
25693-1-AP-20UL |
20 μL |
145.00 € 129.00 € / 20µL Save 16.00 € 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30236450
|
Proteintech
25693-1-AP-150UL |
150 μL |
410.00 € 349.00 € / 150µL Save 61.00 € 14% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ATAD4 / PRR15L antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human ATAD4 / PRR15L. This antibody recognizes human, mouse antigen. The ATAD4 / PRR15L antibody has been validated for the following applications: IHC, IF/ICC, ELISA analysis.Specifications
| ATAD4 / PRR15L | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| MGC11242 | |
| IgG | |
| Antigen affinity purification | |
| Polyclonal Antibody |
| 650 μg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| 79170 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title