missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATAD2 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23894-1-AP-20UL
This item is not returnable.
View return policy
Description
ATAD2 is a member of the ATPase family of proteins. Members of this family share a conserved region of around 220 amino acids that contain an ATP-binding site. They also contain one or two AAA (ATPases Associated with Diverse Cellular Activities) domains. ATPase family proteins perform chaperone-like functions that assist in assembly, operation, and disassembly of protein complexes. Involved in the estrogen-induced cell proliferation and cell cycle progression of breast cancer cells.Specifications
| ATAD2 | |
| Polyclonal | |
| Unconjugated | |
| ATAD2 | |
| ANCCA, ATAD2, PRO2000 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 29028 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunocytochemistry, Western Blot, Immunofluorescence | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6PL18 | |
| ATAD2 | |
| ATAD2 Fusion Protein Ag20947 | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction