missing translation for 'onlineSavingsMsg'
Learn More

AT6, MedChemExpress

Product Code. 16487337
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16487337

Brand: Medchem Express HY112375

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AT6 is a PROTAC AT1 analogue, which is a PROTAC connected by ligands for von Hippel-Lindau and BRD4 with highly selectivity to bromodomain (Brd4).

AT6 is a PROTAC AT1 analogue, which is a PROTAC connected by ligands for von Hippel-Lindau and BRD4 with highly selectivity to bromodomain (Brd4).

Specifications

Chemical Name or Material AT6
CAS 2098836-50-9
Color White
Purity Grade Notes Research
Molecular Formula C48H58ClN9O7S3
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (99.53 mM; Need ultrasonic)
SMILES CC1=C(C)C(C(C2=CC=C(Cl)C=C2)=N[C@@H](CC(NCCOCCOCCSC(C)(C)[C@H](NC(C)=O)C(N3C[C@H](O)C[C@H]3C(NCC4=CC=C(C5=C(C)N=CS5)C=C4)=O)=O)=O)C6=NN=C(C)N67)=C7S1
Molecular Weight (g/mol) 1004.68
Formula Weight 1004.68
Percent Purity 98.74%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.