missing translation for 'onlineSavingsMsg'
Learn More

Astryl, TRC

Product Code. 30397699
Change view
Click to view available options
Quantity:
1 mg
2 mg
10 mg
Unit Size:
10mg
1mg
2mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30397699 10 mg 10mg
30396118 2 mg 2mg
30408686 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397699 Supplier Toronto Research Chemicals Supplier No. TRCH95457510MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 144-87-6
Molecular Formula C8H10NO5As
Molecular Weight (g/mol) 275.09
Synonym Glycarsamide, Glycarsiamidon, N-Glycoloylarsanilic Acid, NSC 163966, NSC 221701, [4-(2-Hydroxyacetamido)phenyl]arsonic Acid, p-(Glycoloylamino)benzenearsonic Acid, p-(Glycoloylamino)phenylarsonic Acid, p-(Hydroxyacetamido)phenylarsonic Acid
IUPAC Name [4-[(2-hydroxyacetyl)amino]phenyl]arsonic acid
SMILES OCC(=O)Nc1ccc(cc1)[As](=O)(O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C8H10AsNO5/c11-5-8(12)10-7-3-1-6(2-4-7)9(13,14)15/h1-4,11H,5H2,(H,10,12)(H2,13,14,15)
Formula Weight 274.9775
Chemical Name or Material Astryl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.