Learn More
Astryl, TRC
Product Code. 30408686
Change view
Click to view available options
Quantity:
1 mg
2 mg
10 mg
Unit Size:
10mg
1mg
2mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30408686 1 mg 1mg
30397699 10 mg 10mg
30396118 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30408686 Supplier Toronto Research Chemicals Supplier No. TRCH9545751MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 144-87-6
Molecular Formula C8H10NO5As
Molecular Weight (g/mol) 275.09
Synonym Glycarsamide, Glycarsiamidon, N-Glycoloylarsanilic Acid, NSC 163966, NSC 221701, [4-(2-Hydroxyacetamido)phenyl]arsonic Acid, p-(Glycoloylamino)benzenearsonic Acid, p-(Glycoloylamino)phenylarsonic Acid, p-(Hydroxyacetamido)phenylarsonic Acid
IUPAC Name [4-[(2-hydroxyacetyl)amino]phenyl]arsonic acid
SMILES OCC(=O)Nc1ccc(cc1)[As](=O)(O)O
Specifications
Quantity 1 mg
InChI Formula InChI=1S/C8H10AsNO5/c11-5-8(12)10-7-3-1-6(2-4-7)9(13,14)15/h1-4,11H,5H2,(H,10,12)(H2,13,14,15)
Formula Weight 274.9775
Chemical Name or Material Astryl