missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASTN2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17351-1-AP-20UL
This item is not returnable.
View return policy
Description
This gene encodes a protein that is expressed in the brain and may function in neuronal migration, based on functional studies of the related astrotactin 1 gene in human and mouse. A deletion at this locus has been associated with schizophrenia. Multiple transcript variants encoding different proteins have been found for this locus.Specifications
| ASTN2 | |
| Polyclonal | |
| Unconjugated | |
| ASTN2 | |
| ASTN2, astrotactin 2, KIAA0634 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 100361323, 23245, 56079 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.21 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O75129, Q80Z10 | |
| Astn2 | |
| ASTN2 Fusion Protein Ag11425 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction