Learn More
Gilson™ ASPEC™ DVB SPE Cartridge, Bed Wt. 100 mg
Polymer based DVB cartridges contain a high-quality, hydrophobic sorbent from polystyrene-divinylbenzene copolymer, high hydrophobicity, for use as a reversed-phase for extraction of neutral, acidic and basic compounds in biological and viscous matrices
Brand:  Gilson™ 54350850
Product Code. 16626605
  • 247.00 € / Pack of 30
Please to purchase this item. Need a web account? Register with us today!
This item is not returnable. View return policy
Description
Description
  • Pre-capped cartridges are ASPEC™ instrument-ready
  • Automation delivers consistent flow rates
    • Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
    • Cartridges available uncapped for manual extractions
Specifications
Specifications
DVB
100 mg
DVB
6 mL
SPE
ASPEC™
SPE cartridge
Yes