Learn More
Gilson™ ASPEC™ DVB SPE Cartridge, Bed Wt. 100 mg
Polymer based DVB cartridges contain a high-quality, hydrophobic sorbent from polystyrene-divinylbenzene copolymer, high hydrophobicity, for use as a reversed-phase for extraction of neutral, acidic and basic compounds in biological and viscous matrices
Brand: Gilson™ 54350850
Description
- Pre-capped cartridges are ASPEC™ instrument-ready
- Automation delivers consistent flow rates
- Provide loading capacity, accurate elution properties and reproducible separation with low solvent usage
- Cartridges available uncapped for manual extractions
Specifications
| DVB | |
| 100 mg | |
| DVB | |
| 6 mL |
| SPE | |
| ASPEC™ | |
| SPE cartridge | |
| Yes |