missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aspartate beta hydroxylase Antibody (3G5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00000444-M09
This item is not returnable.
View return policy
Description
Aspartate beta hydroxylase Monoclonal antibody specifically detects Aspartate beta hydroxylase in Human samples. It is validated for Western Blot, ELISA, ELISA
Specifications
| Aspartate beta hydroxylase | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| AAH, ASP beta-hydroxylase, aspartate beta-hydroxylaseA beta H-J-J, aspartyl/asparaginyl-beta-hydroxylase, BAH, cardiac junctin, CASQ2BP1, EC 1.14.11.16, HAAHaspartyl/asparaginyl beta-hydroxylase, humbug, JCTN, junctate, junctin, Peptide-aspartate beta-dioxygenase | |
| ASPH (NP_115857, 161 a.a. ~ 270 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. PTGEPQQEDDEFLMATDVDDRFETLEPEVSHEETEHSYHVEETVSQDCNQDMEEMMSEQENPDSSEPVVEDERLHHDTDDVTYQVYEEQAVYEPLENEGIEITEVTAPPE | |
| 0.1 mg | |
| Cancer | |
| 444 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA, Sandwich ELISA | |
| 3G5 | |
| Western Blot 1:500, ELISA, Sandwich ELISA | |
| NP_115857 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction