missing translation for 'onlineSavingsMsg'
Learn More

Asimadoline, MedChemExpress

Product Code. 16484118
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16484118

Brand: Medchem Express HY107384

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

MedChemExpress Asimadoline (EMD-61753) is an orally active, selective and peripherally active κ-opioid agonist with IC50s of 5.6 nM (guinea pig) and 1.2 nM (human recombinant). Asimadoline has low permeability across the blood brain barrier and has peripheral anti-inflammatory actions. Asimadoline ameliorates allodynia in diabetic rats and has the potential for irritable bowel syndrome (IBS).

Asimadoline (EMD-61753) is an orally active, selective and peripherally active κ-opioid agonist with IC50s of 5.6 nM (guinea pig) and 1.2 nM (human recombinant). Asimadoline has low permeability across the blood brain barrier and has peripheral anti-inflammatory actions. Asimadoline ameliorates allodynia in diabetic rats and has the potential for irritable bowel syndrome (IBS).

Specifications

Chemical Name or Material Asimadoline
Physical Form Solid
CAS 153205-46-0
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C27H30N2O2
For Use With (Application) COVID-19-immunoregulation
Synonym EMD-61753
Solubility Information DMSO : ≥ 100 mg/mL (241.23 mM)
SMILES O[C@H]1CCN(C1)C[C@H](C2=CC=CC=C2)N(C)C(C(C3=CC=CC=C3)C4=CC=CC=C4)=O
Molecular Weight (g/mol) 414.54
Formula Weight 414.54
Percent Purity 99.36%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.