Learn More
Asimadoline, MedChemExpress
Product Code. 16486817
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Product Code. 16486817
Brand: Medchem Express HY107384
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.
Request a Price for this item here.
MedChemExpress Asimadoline (EMD-61753) is an orally active, selective and peripherally active κ-opioid agonist with IC50s of 5.6 nM (guinea pig) and 1.2 nM (human recombinant). Asimadoline has low permeability across the blood brain barrier and has peripheral anti-inflammatory actions. Asimadoline ameliorates allodynia in diabetic rats and has the potential for irritable bowel syndrome (IBS).
Asimadoline (EMD-61753) is an orally active, selective and peripherally active κ-opioid agonist with IC50s of 5.6 nM (guinea pig) and 1.2 nM (human recombinant). Asimadoline has low permeability across the blood brain barrier and has peripheral anti-inflammatory actions. Asimadoline ameliorates allodynia in diabetic rats and has the potential for irritable bowel syndrome (IBS).
Specifications
Chemical Name or Material Asimadoline
Physical Form Solid
CAS 153205-46-0
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C27H30N2O2
For Use With (Application) COVID-19-immunoregulation
Synonym EMD-61753
Solubility Information DMSO : ≥ 100 mg/mL (241.23 mM)
SMILES O[C@H]1CCN(C1)C[C@H](C2=CC=CC=C2)N(C)C(C(C3=CC=CC=C3)C4=CC=CC=C4)=O
Molecular Weight (g/mol) 414.54
Formula Weight 414.54
Percent Purity 99.36%
Grade Research