Learn More
ASIC2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17851-1-AP-20UL
This item is not returnable.
View return policy
Description
ACCN1 encodes a member of the degenerin/epithelial sodium channel (DEG/ENaC) superfamily. The members of this family are amiloride-sensitive sodium channels that contain intracellular N and C termini, 2 hydrophobic transmembrane regions, and a large extracellular loop, which has many cysteine residues with conserved spacing. The member encoded by this protein may play a role in neurotransmission. In addition, a heteromeric association between this member and ACCN3 (variant 1) has been observed to co-assemble into proton-gated channels sensitive to gadolinium.Specifications
| ASIC2 | |
| Polyclonal | |
| Unconjugated | |
| ASIC2 | |
| ACCN, ACCN1, Acid sensing ion channel 2, ASIC2, ASIC2a, BNaC1, BNC1, Brain sodium channel 1, hBNaC1, Mammalian degenerin homolog, MDEG | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11418, 25364, 40 | |
| -20°C | |
| Liquid |
| Western Blot, Immunoprecipitation | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q16515, Q62962, Q925H0 | |
| ASIC2 | |
| ASIC2 Fusion Protein Ag12301 | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |