Learn More
ASF/SF2 Mouse anti-Human, Mouse, Rat, Clone: 1A6C5, Proteintech
Mouse Monoclonal Antibody
145.00 € - 410.00 €
Specifications
| Antigen | ASF/SF2 |
|---|---|
| Clone | 1A6C5 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16888765
|
Proteintech
66671-1-IG-20UL |
20 μL |
145.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16878765
|
Proteintech
66671-1-IG-150UL |
150 μL |
410.00 €
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This antibody can recognize all the 3 isoforms (28 kDa,33 kDa and 22 kDa) of SFRS1.
UBE2E2 catalyzes the covalent attachment of ubiquitin to other proteins.Specifications
| ASF/SF2 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Alternative splicing factor 1, ASF, ASF 1, ASF/SF2, SF2, SF2p33, SFRS1, SRp30a, SRSF1 | |
| SRSF1 | |
| IgG1 | |
| Protein G | |
| Antibody |
| 1A6C5 | |
| Western Blot, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Mouse, Human, Rat | |
| Q07955, Q6PDM2 | |
| 110809, 6426, 689890 | |
| ASF/SF2 Fusion Protein Ag3626 | |
| Primary | |
| -20°C | |
| SRSF1 |