missing translation for 'onlineSavingsMsg'
Learn More

ASCL3 Antibody, Novus Biologicals™

Product Code. 18254542 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18254542 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18254542 Supplier Novus Biologicals Supplier No. NBP168871

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

ASCL3 Polyclonal specifically detects ASCL3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ASCL3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.1 ug/ml, Immunohistochemistry 0.1-0.3ug/ml, Immunohistochemistry-Paraffin, Peptide ELISA detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_064435.1
Gene Alias achaete-scute complex homolog 3 (Drosophila), ASH-3, bHLH transcription factor Sgn-1 (Salivary Glands 1), bHLH transcriptional regulator Sgn-1, BHLHA42, bHLHa42achaete-scute homolog 3, Class A basic helix-loop-helix protein 42, hASH3, HASH3achaete-scute complex (Drosophila) homolog-like 3, SGN1
Gene Symbols ASCL3
Host Species Goat
Immunogen Peptide with sequence C-HLPEDYLEKRLSK corresponding to internal region according to Uniprot Mouse NP_064435.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56676
Test Specificity Expected from sequence similarity: Cow.
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.