missing translation for 'onlineSavingsMsg'
Learn More

Asatone, MedChemExpress

Product Code. 16491758
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16491758

Brand: Medchem Express HYN6826

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Asatone is an active component isolated from Radix et Rhizoma Asari, with anti-inflammatory effect via activation of NF-κB and donwn regulation of p-MAPK (ERK, JNK and p38) pathways.

Asatone is an active component isolated from Radix et Rhizoma Asari, with anti-inflammatory effect via activation of NF-κB and donwn regulation of p-MAPK (ERK, JNK and p38) pathways.

Specifications

Chemical Name or Material Asatone
CAS 38451-63-7
Color White
Purity Grade Notes Research
Molecular Formula C24H32O8
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (222.96 mM; Need ultrasonic)
SMILES O=C1C(OC)(OC)[C@@]2([H])[C@@](C(OC)(OC)C3=O)([H])C(CC=C)=C[C@@]3(OC)[C@@]2(CC=C)C=C1OC
Molecular Weight (g/mol) 448.51
Formula Weight 448.51
Percent Purity 99.88%
Grade Research
Physical Form Crystals
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.