missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARTS Monoclonal Antibody (ARTS51)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA124693
This item is not returnable.
View return policy
Description
MA1-24693 is expected to cross react with mouse (100% conserved) due to sequence homology. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Apoptosis is related to many diseases and development. Mitochondrial proteins, such as cytochrome c, Apaf-1, and AIF play important role in apoptosis. A novel mitochondrial septin-like protein was identified recently and designated ARTS for apoptosis related protein in TGF-beta signaling pathway. ARTS that is encoded by the human septin H5/PNUTL2/CDCrel2b gene is located to mitochondria and translocates to the nucleus when apoptosis occurs. ARTS is expressed in many tissues. It enhances cell death induced by TGF-beta and, to a lesser extent, by other apoptotic agents, such as TNF-a and Fas ligand.
Specifications
| ARTS | |
| Monoclonal | |
| 2.2 mg/mL | |
| PBS with 15mM sodium azide; pH 7.4 | |
| O43236 | |
| SEPTIN4 | |
| Synthetic peptide corresponding to C-terminal region (amino acids 247-266) of human ARTS protein, conjugated to KLH. | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| ARTS51 | |
| Unconjugated | |
| SEPTIN4 | |
| 4-Sep; apoptosis-related protein in the TGF-beta signaling pathway; ARTS; Bh5; BRADEION; Bradeion beta; brain protein H5; C17orf47; CE5B3; CE5B3 beta; cell division control-related protein 2; cell division control-related protein 2b; cerebral protein 7; EG3-1RVC; EG3RVC; expression gene 3 in rat visual cortex; expression gene 3-1 in rat visual cortex; H5; hCDCREL-2; hucep-7; MART; M-Septin; OTTMUSG00000001265; peanut-like 2; peanut-like 2 homolog; peanut-like protein 2; PNUTL2; SEP4; Sept4; septin 4; septin H5; Septin4; septin-4; septin-M; Uncharacterized protein C17orf47 | |
| Mouse | |
| Purified | |
| RUO | |
| 5414 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction