missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Artemis Recombinant Rabbit Monoclonal Antibody (HL1227)
GREENER_CHOICE

Product Code. 17859555
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17859555 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17859555 Supplier Invitrogen™ Supplier No. MA546962

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial.

The protein encoded by this gene is a receptor for aryl hydrocarbons and a ligand-activated transcription factor. The encoded protein is found in the cytoplasm as part of a multiprotein complex, but upon binding of ligand is transported to the nucleus. This protein can regulate the expression of many xenobiotic metabolizing enzymes. Also, the encoded protein can bind specifically to and inhibit the activity of hepatitis B virus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Artemis
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL1227
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene DCLRE1C
Gene Accession No. Q96SD1
Gene Alias 9930121L06Rik; AI661365; Art; ARTEMIS; ASCID; A-SCID; Dclre1c; DCLRE1C protein; DCLREC1C; DNA cross-link repair 1A, PSO2 homolog; DNA cross-link repair 1C; DNA cross-link repair 1C (PSO2 homolog, S. cerevisiae); DNA cross-link repair 1C protein; DNA cross-link repair 1C, PSO2 homolog (S. cerevisiae); FLJ11360; hSNM1C; mArt; Protein artemis; Protein A-SCID; RS SCID; RS-SCID; SCIDA; severe combined immunodeficiency, type a (Athabascan); SNM1 homolog C; SNM1C; SNM1C antibody; Snm1l; SNM1-like protein; tein
Gene Symbols DCLRE1C
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Artemis. The exact sequence is proprietary.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64421
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.