missing translation for 'onlineSavingsMsg'
Learn More

ARN2966, MedChemExpress

Product Code. p-200101308
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16447456 100 mg 100mg
16443517 50 mg 50mg
16439977 5 mg 5mg
16417498 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16447456 Supplier Medchem Express Supplier No. HY18292

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ARN2966 is a potent post-transcriptional modulator of APP expression; reduces expression of APP with resultant lower production of A(beta).

ARN2966 is a potent post-transcriptional modulator of APP expression

Specifications

Chemical Name or Material ARN2966
CAS 102212-26-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C12H12N2O
Quantity 100 mg
Solubility Information DMSO : 110 mg/mL (549.34 mM; Need ultrasonic)
SMILES OC1=CC=CC=C1NCC2=NC=CC=C2
Molecular Weight (g/mol) 200.24
Formula Weight 200.24
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.