Learn More
ARN2966, MedChemExpress
Description
ARN2966 is a potent post-transcriptional modulator of APP expression
Specifications
Specifications
| Chemical Name or Material | ARN2966 |
| CAS | 102212-26-0 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C12H12N2O |
| Quantity | 100 mg |
| Solubility Information | DMSO : 110 mg/mL (549.34 mM; Need ultrasonic) |
| SMILES | OC1=CC=CC=C1NCC2=NC=CC=C2 |
| Molecular Weight (g/mol) | 200.24 |
| Formula Weight | 200.24 |
| Show More |