missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ARID5A Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-79440
This item is not returnable.
View return policy
Description
ARID5A Polyclonal specifically detects ARID5A in Human samples. It is validated for Western Blot.
Specifications
| ARID5A | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| ARID domain-containing protein 5A, AT rich interactive domain 5A (MRF1-like), AT-rich interactive domain-containing protein 5A, RFVG5814 | |
| Rabbit | |
| 64 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_997646 | |
| ARID5A | |
| Synthetic peptide directed towards the N terminal of human ARID5AThe immunogen for this antibody is ARID5A. Peptide sequence LWKNVYDELGGSPGSTSAATCTRRHYERLVLPYVRHLKGEDDKPLPTSKP. | |
| Affinity purified | |
| RUO | |
| 10865 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction