missing translation for 'onlineSavingsMsg'
Learn More

ARHGEF17, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16070347
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16070347 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16070347 Supplier Abnova Supplier No. PAB13013.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Specifications

Antigen ARHGEF17
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene Rho guanine nucleotide exchange factor (GEF) 17
Gene Alias FLJ90019/KIAA0337/P164RHOGEF/TEM4/p164-RhoGEF
Gene Symbols ARHGEF17
Host Species Rabbit
Immunogen A synthetic peptide corresponding to internal region 18 amino acids of human ARHGEF17.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9828
Target Species Human
Content And Storage Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.