Learn More
MedChemExpress Argireline
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Purity | 99.0% |
| CAS | 616204-22-9 |
| Content And Storage | Powder -80°C 2 years, -20°C 1 year∣In solvent -80°C 6 months, -20°C 1 month |
| Molecular Weight (g/mol) | 888.99 g/mol |
| Product Type | Peptides |
| Quantity | 50 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 100 mg/mL (112.49 mM∣Need ultrasonic) |
| Synonym | Acetyl hexapeptide-3 |