Learn More
Tocris Bioscience™ Arformoterol Tartrate
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Arformoterol tartrate is a long-acting β2 adrenergic agonist (LABA). Bronchodilator.
Specifications
Specifications
| CAS | 200815-49-2 |
| Quantity | 50 mg |
| Target | Adrenergic beta-2 |
| Molecular Formula | C19H24N2O4.C4H6O6 |
| Formula Weight | 494.49g/mol |
| Product Type | Long-acting Agonist (LABA) |