missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARF7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113182
This item is not returnable.
View return policy
Description
ARF7 Polyclonal Antibody for Western Blot, IHC (P), ELISA
GTPase that recruits MYO1E to MHC class II-containing vesicles via the effector protein ARL14EP and hence controls the movement of these vesicles along the actin cytoskeleton in dendritic cells.
Specifications
| ARF7 | |
| Polyclonal | |
| Unconjugated | |
| ARL14 | |
| 9130014L17Rik; ADP ribosylation factor like GTPase 14; ADP-ribosylation factor 7; ADP-ribosylation factor like GTPase 14; ADP-ribosylation factor-like 14; ADP-ribosylation factor-like protein 14; Arf7; Arl14 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 80117 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 2.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8N4G2 | |
| ARL14 | |
| Recombinant Human ADP-ribosylation factor-like protein 14 protein (2-192AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction